missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbiphenyl, 98%

Catalog No. AAL0131203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0131203 1 g
AAL0131206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0131203 Supplier Thermo Scientific Chemicals Supplier No. L0131203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 643-58-3 | C13H12 | 168.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 643-58-3
Molecular Formula C13H12
Molecular Weight (g/mol) 168.239
MDL Number MFCD00008517
InChI Key ALLIZEAXNXSFGD-UHFFFAOYSA-N
Synonym 2-methylbiphenyl, 2-methyl-1,1'-biphenyl, 2-phenyltoluene, 2-methyl biphenyl, o-phenyltoluene, o-methylbiphenyl, 2-methyl-biphenyl, 1,1'-biphenyl, 2-methyl, biphenyl, 2-methyl, unii-x937mq8s1t
PubChem CID 12563
IUPAC Name 1-methyl-2-phenylbenzene
SMILES CC1=CC=CC=C1C2=CC=CC=C2

Specifications

Melting Point 0°C
Density 1.01
Boiling Point 256°C
Flash Point 107°C (224°F)
Refractive Index 1.591
Quantity 1 g
Beilstein 2039655
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material 2-Methylbiphenyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-400-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only