missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzoxazole 99.0+%, TCI America™

Catalog No. M055725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M055725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M055725G Supplier TCI America Supplier No. M055725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 95-21-6
Boiling Point 201°C
Molecular Formula C8H7NO
Quantity 25 g
MDL Number MFCD00005771
Synonym 2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti
InChI Key DQSHFKPKFISSNM-UHFFFAOYSA-N
SMILES CC1=NC2=CC=CC=C2O1
IUPAC Name 2-methyl-1,3-benzoxazole
Molecular Weight (g/mol) 133.15
PubChem CID 7225
ChEBI CHEBI:51603
Formula Weight 133.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbenzoxazole
Show More Show Less
EINECSNumber (5)-0156
RTECSNumber DM4850000
TSCA Yes