missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzoxazole, 97%

Catalog No. p-7021032
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1319814 25 g
AAA1319822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1319814 Supplier Thermo Scientific Chemicals Supplier No. A1319814

CAS: 95-21-6 | C8H7NO | 133.15 g/mol

2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

Solubility
Not miscible in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 95-21-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005771
InChI Key DQSHFKPKFISSNM-UHFFFAOYSA-N
Synonym 2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3ti
PubChem CID 7225
ChEBI CHEBI:51603
IUPAC Name 2-methyl-1,3-benzoxazole
SMILES CC1=NC2=CC=CC=C2O1

Specifications

Melting Point 2°C to 4°C
Density 1.12
Boiling Point 200°C to 201°C
Flash Point 85°C (185°F)
Refractive Index 1.55
Quantity 25 g
Beilstein 112297
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 2-Methylbenzoxazole
Show More Show Less
Hazard Category H227-H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H301-H318-H227-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-399-9
RTECSNumber DM4850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only