missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzofuran, 96%

Catalog No. AC208400010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208400010 1 g Glass bottle
AC208400050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC208400010 Supplier Thermo Scientific Chemicals Supplier No. 208400010

CAS: 4265-25-2 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4265-25-2
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00005850
InChI Key GBGPVUAOTCNZPT-UHFFFAOYSA-N
Synonym 2-methylbenzofuran, benzofuran, 2-methyl, 2-methylcumarone, 2-methylbenzo b furan, 2-methyl benzofuran, unii-8x3183bz3x, methylbenzofuran, methyl benzoxole, 2-methyl-benzofuran, #
PubChem CID 20263
IUPAC Name 2-methyl-1-benzofuran
SMILES CC1=CC2=CC=CC=C2O1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0570g/mL
Boiling Point 197.0°C to 198.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5618
Quantity 1 g
Beilstein 17,6
Specific Gravity 1.057
Formula Weight 132.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 224-249-1

RUO – Research Use Only