missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzo[b]thiophene 98.0+%, TCI America™

Catalog No. p-7132726
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14291G 1 g
M14295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14291G Supplier TCI America Supplier No. M14291G
Only null left

Chemical Identifiers

CAS 1195-14-8
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00216250
InChI Key BLZKSRBAQDZAIX-UHFFFAOYSA-N
PubChem CID 70952
IUPAC Name 2-methyl-1-benzothiophene
SMILES CC1=CC2=CC=CC=C2S1

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 1 g
Synonym 2-methylbenzo b thiophene, 2-methylthianaphthene, 2-methyl-1-thiaindene, 2-methylbenzothiophene, 2-methylthioindene, benzo b thiophene, 2-methyl, benzo b thiophene,2-methyl, acmc-1bscn, benzothiophene, 2-methyl
Formula Weight 148.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylbenzo[b]thiophene