missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbenzimidazole 98.0+%, TCI America™
Supplier: TCI America M0286250G
Specifications
| 2-Methylbenzimidazole | |
| 175°C | |
| C8H8N2 | |
| 250 g | |
| LDZYRENCLPUXAX-UHFFFAOYSA-N | |
| 2-methyl-1H-1,3-benzodiazole | |
| 11984 | |
| ≥98.0% (GC,T) |
| 615-15-6 | |
| White-Yellow | |
| MFCD00005598 | |
| 2-methylbenzimidazole, 2-methyl-1h-benzo d imidazole, 1h-benzimidazole, 2-methyl, benzimidazole, 2-methyl, 2-methyl-1h-benzoimidazole, 2-methyl-1h-1,3-benzodiazole, acetamidine, n-n'-o-phenylene, 2-methyl benzimidazole, unii-zh8iww7y8b, 2-methylbenaimidazole | |
| CC1=NC2=CC=CC=C2N1 | |
| 132.17 | |
| 132.17 | |
| Crystalline Powder |
Chemical Identifiers
| 615-15-6 | |
| 132.17 | |
| LDZYRENCLPUXAX-UHFFFAOYSA-N | |
| 11984 | |
| CC1=NC2=CC=CC=C2N1 |
| C8H8N2 | |
| MFCD00005598 | |
| 2-methylbenzimidazole, 2-methyl-1h-benzo d imidazole, 1h-benzimidazole, 2-methyl, benzimidazole, 2-methyl, 2-methyl-1h-benzoimidazole, 2-methyl-1h-1,3-benzodiazole, acetamidine, n-n'-o-phenylene, 2-methyl benzimidazole, unii-zh8iww7y8b, 2-methylbenaimidazole | |
| 2-methyl-1H-1,3-benzodiazole |
Safety and Handling
RTECSNumber : DD9100000
TSCA : Yes