missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzimidazole 98.0+%, TCI America™

Catalog No. M028625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M028625G 25 g
M0286250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M028625G Supplier TCI America Supplier No. M028625G

Chemical Identifiers

CAS 615-15-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005598
InChI Key LDZYRENCLPUXAX-UHFFFAOYSA-N
Synonym 2-methylbenzimidazole, 2-methyl-1h-benzo d imidazole, 1h-benzimidazole, 2-methyl, benzimidazole, 2-methyl, 2-methyl-1h-benzoimidazole, 2-methyl-1h-1,3-benzodiazole, acetamidine, n-n'-o-phenylene, 2-methyl benzimidazole, unii-zh8iww7y8b, 2-methylbenaimidazole
PubChem CID 11984
IUPAC Name 2-methyl-1H-1,3-benzodiazole
SMILES CC1=NC2=CC=CC=C2N1

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 25 g
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylbenzimidazole
RTECSNumber DD9100000
TSCA Yes