missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzimidazole, 98%

Catalog No. AAA1729030
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1729030 250 g
AAA1729018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1729030 Supplier Thermo Scientific Chemicals Supplier No. A1729030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 615-15-6 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-15-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005598
InChI Key LDZYRENCLPUXAX-UHFFFAOYSA-N
PubChem CID 11984
SMILES CC1=NC2=CC=CC=C2N1

Specifications

Melting Point 174°C to 178°C
Quantity 250 g
Beilstein 112264
Synonym 2-methylbenzimidazole, 2-methyl-1h-benzo d imidazole, 1h-benzimidazole, 2-methyl, benzimidazole, 2-methyl, 2-methyl-1h-benzoimidazole, 2-methyl-1h-1,3-benzodiazole, acetamidine, n-n'-o-phenylene, 2-methyl benzimidazole, unii-zh8iww7y8b, 2-methylbenaimidazole
IUPAC Name 2-methyl-1H-benzimidazole
Formula Weight 132.17
Percent Purity 98%
Chemical Name or Material 2-Methylbenzimidazole
Hazard Category H302-H315-H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-411-9
RTECSNumber DD9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only