missing translation for 'onlineSavingsMsg'
Learn More

2-Methylanthraquinone, 97%

Catalog No. AAA1450218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1450218 50 g
AAA1450230 250 g
AAA145020B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1450218 Supplier Thermo Scientific Chemicals Supplier No. A1450218
Only null left

CAS: 84-54-8 | C15H10O2 | 222.243 g/mol

2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

Solubility
Soluble in water 1.234 mg/L @ 25°C.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 84-54-8
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00001235
InChI Key NJWGQARXZDRHCD-UHFFFAOYSA-N
Synonym 2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
PubChem CID 6773
ChEBI CHEBI:9427
IUPAC Name 2-methylanthracene-9,10-dione
SMILES CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 170°C to 174°C
Flash Point 209°C (408°F)
Quantity 50 g
Beilstein 2050523
Merck Index 14,6021
Solubility Information Soluble in water 1.234mg/L @ 25°C.
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material 2-Methylanthraquinone
EINECSNumber 201-539-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only