missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester, 96%

Catalog No. p-7036395
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54682MD 250 mg
AAH5468203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54682MD Supplier Thermo Scientific Chemicals Supplier No. H54682MD
Only null left

CAS: 904326-88-1 | C11H18BN3O2 | 235.094 g/mol

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 904326-88-1
Molecular Formula C11H18BN3O2
Molecular Weight (g/mol) 235.094
MDL Number MFCD11878345
InChI Key QDOXNCAIXITTKA-UHFFFAOYSA-N
Synonym n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid, pinacol ester, n-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid,pinacol ester, 2-methylamino pyrimidine-5-boronic acid pinacol ester, 2-methylamino pyrimidin-5-yl boronic acid pinacol ester, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl-amine, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl amine, n-m ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, n-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-amine
PubChem CID 46739641
IUPAC Name N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC

Specifications

Melting Point 122°C to 125°C
Quantity 250 mg
Formula Weight 235.09
Percent Purity 96%
Chemical Name or Material 2-(Methylamino)pyrimidine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only