missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)isobutyric Acid Hydrate 98.0+%, TCI America™

Catalog No. M13881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13881G 1 g
M13885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13881G Supplier TCI America Supplier No. M13881G

Chemical Identifiers

CAS 2566-34-9
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00004191
InChI Key DLAMVQGYEVKIRE-UHFFFAOYSA-N
Synonym 2-methylamino isobutyric acid, n,2-dimethylalanine, 2-methyl-2-methylamino propanoic acid, alpha-methylamino isobutyric acid, meaib, alanine, n,2-dimethyl, n-methyl-a-aminoisobutyric acid, 2-methylamino-2-methylpropionic acid, alpha-methylamino-isobutyric acid, 2,n-dimethylalanine
PubChem CID 75725
IUPAC Name 2-methyl-2-(methylamino)propanoic acid
SMILES CC(C)(C(=O)O)NC

Specifications

Color White
Quantity 1 g
Formula Weight 117.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylamino)isobutyric Acid Hydrate