missing translation for 'onlineSavingsMsg'
Learn More

2-Methylallylamine 98.0+%, TCI America™

Catalog No. p-7133391
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
5 mL
Catalog No. Quantity
M27261ML 1 mL
M27265ML 5 mL
2 options

Catalog No. M27261ML

Supplier: TCI America M27261ML

Only null left

Chemical Identifiers

CAS 2878-14-0
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD00053646
InChI Key VXDHQYLFEYUMFY-UHFFFAOYSA-N
Synonym 2-methylallylamine, methallylamine, 2-methylprop-2-en-1-ylamine, 2-propen-1-amine, 2-methyl, unii-fhj35we6dg, fhj35we6dg, 2-methyl-allylamine, 2-methyl-2-propenylamine, 2-methyl-2-propen-1-amine, ksc493c1t
PubChem CID 76141
IUPAC Name 2-methylprop-2-en-1-amine
SMILES CC(=C)CN

Specifications

Color Yellow
Boiling Point 81°C
Quantity 1 mL
UN Number 2733
Formula Weight 71.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methylallylamine