missing translation for 'onlineSavingsMsg'
Learn More

2-Methylallylamine, 97%

Catalog No. AC330700010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330700010 1 g Glass bottle
AC330700050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330700010 Supplier Thermo Scientific Chemicals Supplier No. 330700010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2878-14-0 | C4H9N | 71.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2878-14-0
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD00053646
InChI Key VXDHQYLFEYUMFY-UHFFFAOYSA-N
Synonym 2-methylallylamine, methallylamine, 2-methylprop-2-en-1-ylamine, 2-propen-1-amine, 2-methyl, unii-fhj35we6dg, fhj35we6dg, 2-methyl-allylamine, 2-methyl-2-propenylamine, 2-methyl-2-propen-1-amine, ksc493c1t
PubChem CID 76141
IUPAC Name 2-methylprop-2-en-1-amine
SMILES CC(=C)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.7800g/mL
Boiling Point 78°C
Flash Point -28°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4290 to 1.432
Linear Formula CH3CCH2CH2NH2
Quantity 1 g
Specific Gravity 0.78
Formula Weight 71.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methylallylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-724-2
TSCA TSCA

RUO – Research Use Only