missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Methylacetophenone 98.0+%, TCI America™
$100.67 - $950.67
Chemical Identifiers
| CAS | 577-16-2 |
|---|---|
| Molecular Formula | C9H10O |
| Molecular Weight (g/mol) | 134.178 |
| MDL Number | MFCD00008734 |
| InChI Key | YXWWHNCQZBVZPV-UHFFFAOYSA-N |
| Synonym | 2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl |
| PubChem CID | 11340 |
| IUPAC Name | 1-(2-methylphenyl)ethanone |
| SMILES | CC1=CC=CC=C1C(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M017225ML
|
TCI America
M017225ML |
25 mL |
Each for $100.67
|
|
|||||
|
M0172500ML
|
TCI America
M0172500ML |
500 mL |
Each for $950.67
|
|
|||||
Chemical Identifiers
| 577-16-2 | |
| 134.178 | |
| YXWWHNCQZBVZPV-UHFFFAOYSA-N | |
| 11340 | |
| CC1=CC=CC=C1C(=O)C |
| C9H10O | |
| MFCD00008734 | |
| 2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl | |
| 1-(2-methylphenyl)ethanone |
Specifications
| 577-16-2 | |
| 216°C | |
| MFCD00008734 | |
| 2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl | |
| CC1=CC=CC=C1C(=O)C | |
| 134.178 | |
| 134.18 | |
| Liquid |
| Yellow | |
| C9H10O | |
| 25 mL | |
| YXWWHNCQZBVZPV-UHFFFAOYSA-N | |
| 1-(2-methylphenyl)ethanone | |
| 11340 | |
| ≥98.0% (GC) | |
| 2′-Methylacetophenone |
Safety and Handling
EINECSNumber : (3)-1239
TSCA : No