missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetophenone, 98%

Catalog No. AAA1169522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1169522 100 g
AAA1169506 5 g
AAA1169514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1169522 Supplier Thermo Scientific Chemicals Supplier No. A1169522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 577-16-2 | C9H10O | 134.178 g/mol

2'-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 577-16-2
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00008734
InChI Key YXWWHNCQZBVZPV-UHFFFAOYSA-N
Synonym 2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
PubChem CID 11340
IUPAC Name 1-(2-methylphenyl)ethanone
SMILES CC1=CC=CC=C1C(=O)C

Specifications

Density 1.02
Boiling Point 214°C
Flash Point 75°C (167°F)
Refractive Index 1.5318
Quantity 100 g
Beilstein 907005
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 2'-Methylacetophenone
Hazard Category H227-H312-H332
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P261-P271-P280-P302+P352-P304+P340-P312-P363-P370+P378q-P501c
EINECSNumber 209-408-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only