missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetoacetanilide 98.0+%, TCI America™

Catalog No. A1020500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1020500G 500 g
A102025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1020500G Supplier TCI America Supplier No. A1020500G

Chemical Identifiers

CAS 93-68-5
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00008782
InChI Key TVZIWRMELPWPPR-UHFFFAOYSA-N
Synonym 2'-methylacetoacetanilide, n-2-methylphenyl-3-oxobutanamide, o-acetoacetotoluidide, n-acetoacetyl-o-toluidine, acetoacet-o-toluidide, acetoacet-o-toluidine, 2-acetoacetylaminotoluene, 3-oxo-n-o-tolyl butanamide, butanamide, n-2-methylphenyl-3-oxo, acetoacetyl-2-methylanilide
PubChem CID 7154
IUPAC Name N-(2-methylphenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1C

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 500 g
Formula Weight 191.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Methylacetoacetanilide
EINECSNumber (3)-0204
RTECSNumber AK6550000
TSCA Yes