missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-L-cysteine hydrochloride, 98%, 98% ee, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 441101000

Catalog No. AC441101000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Methyl-L-cysteine hydrochloride
148766-37-4
100.0
Authentic
Glass Bottle
100mg
r-2-amino-3-mercapto-2-methylpropanoic acid hydrochloride, r-2-methylcysteine hcl, r-2-methylcysteine hydrochloride, cysteine, 2-methyl-, hydrochloride, r-2-amino-3-mercapto-2-methyl-propionic acid hydrochloride, 2r-2-amino-2-methyl-3-sulfanyl-propanoic acid hydrochloride, 2r-2-amino-2-methyl-3-sulfanylpropanoic acid hydrochloride, 2r-2-amino-3-mercapto-2-methylpropanoic acid, l-cysteine, 2-methyl-,hydrochloride 1:1, r-l-methylcysteine?hcl
CC(CS)(C(=O)O)N.Cl
171.65
171.65
98% ee
97.5
98.0% min.
98%
C4H9NO2S·ClH
7
MAGCVRLGTQSVGF-WCCKRBBISA-N
(2R)-2-amino-2-methyl-3-sulfanylpropanoic acid;hydrochloride
16679763
98%,98% ee

Chemical Identifiers

148766-37-4
171.65
r-2-amino-3-mercapto-2-methylpropanoic acid hydrochloride, r-2-methylcysteine hcl, r-2-methylcysteine hydrochloride, cysteine, 2-methyl-, hydrochloride, r-2-amino-3-mercapto-2-methyl-propionic acid hydrochloride, 2r-2-amino-2-methyl-3-sulfanyl-propanoic acid hydrochloride, 2r-2-amino-2-methyl-3-sulfanylpropanoic acid hydrochloride, 2r-2-amino-3-mercapto-2-methylpropanoic acid, l-cysteine, 2-methyl-,hydrochloride 1:1, r-l-methylcysteine?hcl
(2R)-2-amino-2-methyl-3-sulfanylpropanoic acid;hydrochloride
C4H9NO2S·ClH
MAGCVRLGTQSVGF-WCCKRBBISA-N
16679763
CC(CS)(C(=O)O)N.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only