missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Methyl-DL-serine hydrate, 96%

Catalog No. AC339750010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC339750010 1 g Glass bottle
AC339750050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC339750010 Supplier Thermo Scientific Chemicals Supplier No. 339750010
Only null left

CAS: 5424-29-3 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5424-29-3
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD03791288
InChI Key CDUUKBXTEOFITR-UHFFFAOYNA-N
Synonym 2-methylserine, alpha-methyl-dl-serine, dl-2-methylserine, alpha-methylserine, 2-methyl-dl-serine, dl-serine, 2-methyl, dl-alpha-methylserine, dl-.alpha.-methylserine, dl-2-methylserine hydrate, .alpha.-methyl-dl-serine
PubChem CID 94309
ChEBI CHEBI:17799
IUPAC Name 2-amino-3-hydroxy-2-methylpropanoic acid
SMILES CC(N)(CO)C(O)=O

Specifications

Melting Point 253.0°C to 263.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids
Formula Weight 119.12
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-DL-serine hydrate, 98%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-565-5
RTECSNumber VT9630200

RUO – Research Use Only