missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-9,10-di(2-naphthyl)anthracene 97.0+%, TCI America™

Catalog No. p-7133452
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
M2821200MG 200 mg
M28211G 1 g
2 options

Catalog No. M2821200MG

Supplier: TCI America M2821200MG

Only null left

Chemical Identifiers

CAS 804560-00-7
Molecular Formula C35H24
Molecular Weight (g/mol) 444.58
MDL Number MFCD12022447
InChI Key HNWFFTUWRIGBNM-UHFFFAOYSA-N
PubChem CID 53403806
IUPAC Name 2-methyl-9,10-bis(naphthalen-2-yl)anthracene
SMILES CC1=CC=C2C(=C1)C(C1=CC=C3C=CC=CC3=C1)=C1C=CC=CC1=C2C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 256°C
Color Yellow
Quantity 200 mg
Formula Weight 444.58
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-9,10-di(2-naphthyl)anthracene