missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Description
Participate in Horner-Wadsworth-Emmons reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Participate in Horner-Wadsworth-Emmons reaction.
Solubility
Sparingly Soluble (6.1E-4 g/L) (25°C).
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications
Specifications
| Melting Point | 45°C to 48°C |
| Quantity | 1 g |
| Sensitivity | Air Sensitive |
| Solubility Information | Sparingly Soluble (6.1E-4g/L) (25°C). |
| Formula Weight | 189.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only