missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)nicotinic acid, Tech., Thermo Scientific™

Catalog No. p-3075441
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AW00680DA 1g Amber Glass Bottle
AW00680EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AW00680DA Supplier Thermo Scientific Chemicals Supplier No. AW00680DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 261635-93-2
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD01311990
InChI Key ODWJFUQRQUCRJZ-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl nicotinic acid, 2-methyl-6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethyl-2-methylpyridine-3-carboxylic acid, 2-methyl-6-trifluoromethyl-nicotinic acid, maybridge4_000139, 2-methyl-6-trifluoromethynicotinic acid, 2-methyl-6-trifluoromethyinicotinic acid, 2-methyl-6-trifluoromethylnicotinic acid, 2-methyl-6-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 2775683
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES CC1=C(C=CC(=N1)C(F)(F)F)C(O)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 205.13
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)nicotinic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.