missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)nicotinic acid, Tech., Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AW00680EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. AW00680EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

261635-93-2
205.14
ODWJFUQRQUCRJZ-UHFFFAOYSA-N
2775683
CC1=C(C=CC(=N1)C(F)(F)F)C(O)=O
C8H6F3NO2
MFCD01311990
2-methyl-6-trifluoromethyl nicotinic acid, 2-methyl-6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethyl-2-methylpyridine-3-carboxylic acid, 2-methyl-6-trifluoromethyl-nicotinic acid, maybridge4_000139, 2-methyl-6-trifluoromethynicotinic acid, 2-methyl-6-trifluoromethyinicotinic acid, 2-methyl-6-trifluoromethylnicotinic acid, 2-methyl-6-trifluoromethyl-3-pyridinecarboxylic acid
2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid

Specifications

2-Methyl-6-(trifluoromethyl)nicotinic acid
80.0
Amber Glass Bottle
10g
2-methyl-6-trifluoromethyl nicotinic acid, 2-methyl-6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethyl-2-methylpyridine-3-carboxylic acid, 2-methyl-6-trifluoromethyl-nicotinic acid, maybridge4_000139, 2-methyl-6-trifluoromethynicotinic acid, 2-methyl-6-trifluoromethyinicotinic acid, 2-methyl-6-trifluoromethylnicotinic acid, 2-methyl-6-trifluoromethyl-3-pyridinecarboxylic acid
CC1=C(C=CC(=N1)C(F)(F)F)C(O)=O
205.14
205.13
Technical
261635-93-2
100.0
C8H6F3NO2
MFCD01311990
ODWJFUQRQUCRJZ-UHFFFAOYSA-N
2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid
2775683
Tech.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents