missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)nicotinic acid, 97%

Catalog No. AAH3300309
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3300309 10 g
AAH3300303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3300309 Supplier Thermo Scientific Chemicals Supplier No. H3300309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 261635-93-2 | C8H6F3NO2 | 205.14 g/mol

2-Methyl-6-(trifluoromethyl)nicotinic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-6-(trifluoromethyl)nicotinic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water (4.7 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 261635-93-2
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD01311990
InChI Key ODWJFUQRQUCRJZ-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl nicotinic acid, 2-methyl-6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethyl-2-methylpyridine-3-carboxylic acid, 2-methyl-6-trifluoromethyl-nicotinic acid, maybridge4_000139, 2-methyl-6-trifluoromethynicotinic acid, 2-methyl-6-trifluoromethyinicotinic acid, 2-methyl-6-trifluoromethylnicotinic acid, 2-methyl-6-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 2775683
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES CC1=C(C=CC(=N1)C(F)(F)F)C(O)=O

Specifications

Melting Point 139°C to 141°C
Quantity 10 g
Solubility Information Slightly soluble in water (4.7g/L at 25°C).
Formula Weight 205.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)nicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only