missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-phenylpyridine 98.0+%, TCI America™
$253.51 - $1008.46
Chemical Identifiers
| CAS | 46181-30-0 |
|---|---|
| Molecular Formula | C12H11N |
| Molecular Weight (g/mol) | 169.23 |
| MDL Number | MFCD00796537 |
| InChI Key | GREMYQDDZRJQEG-UHFFFAOYSA-N |
| PubChem CID | 2762885 |
| IUPAC Name | 2-methyl-6-phenylpyridine |
| SMILES | CC1=CC=CC(=N1)C1=CC=CC=C1 |
Chemical Identifiers
| 46181-30-0 | |
| 169.23 | |
| GREMYQDDZRJQEG-UHFFFAOYSA-N | |
| 2-methyl-6-phenylpyridine |
| C12H11N | |
| MFCD00796537 | |
| 2762885 | |
| CC1=CC=CC(=N1)C1=CC=CC=C1 |
Specifications
| 46181-30-0 | |
| 155°C | |
| MFCD00796537 | |
| GREMYQDDZRJQEG-UHFFFAOYSA-N | |
| 2-methyl-6-phenylpyridine | |
| 2762885 | |
| ≥98.0% (GC,T) | |
| 2-Methyl-6-phenylpyridine |
| Yellow | |
| C12H11N | |
| 1 g | |
| CC1=CC=CC(=N1)C1=CC=CC=C1 | |
| 169.23 | |
| 169.23 | |
| Liquid |
Safety and Handling
TSCA : No