missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroquinoline, 98%

Catalog No. AAA1094503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1094503 1 g
AAA1094506 5 g
AAA1094514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1094503 Supplier Thermo Scientific Chemicals Supplier No. A1094503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 613-30-9 | C10H8N2O2 | 188.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-30-9
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00051736
InChI Key DXDPHHQJZWWAEH-UHFFFAOYSA-N
Synonym 6-nitroquinaldine, quinoline, 2-methyl-6-nitro, maybridge1_001441, acmc-1bb88, 1-2-aminoethyl pyrrole-2,5-dione,hydrochloride
PubChem CID 69172
IUPAC Name 2-methyl-6-nitroquinoline
SMILES CC1=CC=C2C=C(C=CC2=N1)[N+]([O-])=O

Specifications

Melting Point 162°C to 166°C
Quantity 1 g
Beilstein 7915
Formula Weight 188.19
Percent Purity 98%
Chemical Name or Material 2-Methyl-6-nitroquinoline
Hazard Category H302-H315-H319-H335-H351
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only