missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitrobenzoxazole, 98%

Catalog No. p-7026424
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2241506 5 g
AAB2241514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2241506 Supplier Thermo Scientific Chemicals Supplier No. B2241506
Only null left

CAS: 5683-43-2 | C8H6N2O3 | 178.147 g/mol

2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.

Solubility
Soluble in oils. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 5683-43-2
Molecular Formula C8H6N2O3
Molecular Weight (g/mol) 178.147
MDL Number MFCD00022848
InChI Key DPVZLXWUFYMLBK-UHFFFAOYSA-N
PubChem CID 79766
IUPAC Name 2-methyl-6-nitro-1,3-benzoxazole
SMILES CC1=NC2=C(O1)C=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 154°C to 156°C
Quantity 5 g
Synonym 2-methyl-6-nitrobenzoxazole, benzoxazole, 2-methyl-6-nitro, acmc-1azgy, 2-methyl-6-nitro-benzoxazole, benzoxazole,2-methyl-6-nitro, 2-methyl-6-nitrobenzo d oxazole
Solubility Information Soluble in oils. Insoluble in water.
Formula Weight 146.17
Percent Purity 98%
Chemical Name or Material 2-Methyl-6-nitrobenzoxazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-149-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only