missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-propionylfuran 98.0+%, TCI America™

Catalog No. m25075g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25075G 5 g
M250725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25075G Supplier TCI America Supplier No. M25075G
Only null left

Chemical Identifiers

CAS 10599-69-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00053142
InChI Key BXLPZYAVKVFXEO-UHFFFAOYSA-N
Synonym 2-methyl-5-propionylfuran, 1-5-methylfuran-2-yl propan-1-one, 5-methyl-2-propionylfuran, 2-methyl-5-propionyl-furan, 1-5-methyl-2-furyl propan-1-one, 1-propanone,1-5-methyl-2-furanyl, 1-propanone, 1-5-methyl-2-furanyl, 2-methyl-5-propanylfuran, furan, 2-methyl-5-propionyl, furan, 5-methyl-2-propionyl
PubChem CID 82757
IUPAC Name 1-(5-methylfuran-2-yl)propan-1-one
SMILES CCC(=O)C1=CC=C(C)O1

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-5-propionylfuran
TSCA No
Recommended Storage Refrigerator