missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-5-propionylfuran 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 72°C |
| Quantity | 25 g |
| Formula Weight | 138.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Methyl-5-propionylfuran |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |