missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenylbenzoxazole 98.0+%, TCI America™

Catalog No. M124425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M124425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M124425G Supplier TCI America Supplier No. M124425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61931-68-8
Molecular Formula C14H11NO
Molecular Weight (g/mol) 209.248
MDL Number MFCD00005772
InChI Key CHZDPBVCBBQQRP-UHFFFAOYSA-N
Synonym 2-methyl-5-phenylbenzoxazole, 2-methyl-5-phenylbenzo d oxazole, benzoxazole, 2-methyl-5-phenyl, 2-methyl-5-phenyl-benzooxazole, 2-methyl-5-phenyl benzoxazole, pubchem8655, acmc-1b9i5, 2-methyl-5-phenylbenzooxazole, 2-methyl-5-phenyl-benzoxazole, benzoxazole,2-methyl-5-phenyl
PubChem CID 112786
IUPAC Name 2-methyl-5-phenyl-1,3-benzoxazole
SMILES CC1=NC2=C(O1)C=CC(=C2)C3=CC=CC=C3

Specifications

Melting Point 64°C
Boiling Point 158°C
Quantity 25 g
UN Number 2811
Formula Weight 209.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-phenylbenzoxazole
EINECSNumber (5)-0160
RTECSNumber DM4865170
TSCA Yes