missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroimidazole, 99%, Thermo Scientific™

Chemical Identifiers

CAS 88054-22-2
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.1
MDL Number MFCD00151322
InChI Key FFYTTYVSDVWNMY-UHFFFAOYSA-N
Synonym 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
PubChem CID 12760
IUPAC Name 2-methyl-5-nitro-1H-imidazole
SMILES CC1=NC=C(N1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC159470500
SDS View Documents
Thermo Scientific Chemicals
159470500
50g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC159472500
SDS View Documents
Thermo Scientific Chemicals
159472500
250g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

88054-22-2
127.1
FFYTTYVSDVWNMY-UHFFFAOYSA-N
12760
CC1=NC=C(N1)[N+](=O)[O-]
C4H5N3O2
MFCD00151322
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
2-methyl-5-nitro-1H-imidazole

Specifications

88054-22-2
100.0
99%
C4H5N3O2
50g
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
CC1=NC=C(N1)[N+](=O)[O-]
127.1
127.1
2-Methyl-5-nitroimidazole, 99%
98.5
Authentic
Plastic bottle
MFCD00151322
23, I, 23
FFYTTYVSDVWNMY-UHFFFAOYSA-N
2-methyl-5-nitro-1H-imidazole
12760
99%
Videos
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only