missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroimidazole, 99%

Catalog No. AC159472500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159472500 250 g Plastic bottle
AC159470500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159472500 Supplier Thermo Scientific Chemicals Supplier No. 159472500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88054-22-2 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88054-22-2
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00005191
InChI Key FFYTTYVSDVWNMY-UHFFFAOYSA-N
Synonym 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
PubChem CID 12760
IUPAC Name 2-methyl-5-nitro-1H-imidazole
SMILES CC1=NC=C(N1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 252°C to 254°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 250 g
Beilstein 23, I, 23
Formula Weight 127.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Methyl-5-nitroimidazole, 99%
Show More Show Less

RUO – Research Use Only