missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-hexen-3-ol, 97%

Catalog No. AAB2022006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2022006 5 g
AAB2022014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2022006 Supplier Thermo Scientific Chemicals Supplier No. B2022006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32815-70-6 | C7H14O | 114.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32815-70-6
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00048326
InChI Key JKGMJZOZIJHPOH-UHFFFAOYSA-N
PubChem CID 141746
IUPAC Name 2-methylhex-5-en-3-ol
SMILES CC(C)C(CC=C)O

Specifications

Quantity 5 g
UN Number UN1987
Synonym 2-methyl-5-hexen-3-ol, 5-hexen-3-ol, 2-methyl, allyl isopropyl carbinol, acmc-1aev3, 2-methyl-hex-5-en-3-ol, 5-hexen-3-ol,2-methyl
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 2-Methyl-5-hexen-3-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only