missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Catalog No. CC20101CB
Change view
Click to view available options
Quantity:
250mg
1g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC20101CB 250mg
CC20101DA 1g
CC20101EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC20101CB Supplier Thermo Scientific Chemicals Supplier No. CC20101CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127958-10-5
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
MDL Number MFCD02682072
InChI Key XKJAQESOTNACHT-UHFFFAOYSA-N
Synonym 2-methyl-4-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl-4-phenyl, 5-pyrimidinecarboxylic acid, 2-methyl-4-phenyl, acmc-1c0qx, 2-methyl-4-phenylpyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid,2-methyl-4-phenyl, 2-methyl-4-phenyl-pyrimidine-5-carboxylic acid
PubChem CID 2776510
IUPAC Name 2-methyl-4-phenylpyrimidine-5-carboxylic acid
SMILES CC1=NC=C(C(=N1)C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.