missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-phenyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 32002-72-5
Molecular Formula C11H8NO2S
Molecular Weight (g/mol) 218.25
MDL Number MFCD07348569
InChI Key APXSARIYDLVPAY-UHFFFAOYSA-M
Synonym 2-methyl-4-phenylthiazole-5-carboxylic acid, 2-methy-4-phenylthiazol-5-ylcarboxylic acid, 2-methyl-4-phenyl-thiazole-5-carboxylic acid, 2-methyl-4-phenylthiazol-5-ylcarboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl, 5-thiazolecarboxylic acid, 2-methyl-4-phenyl
PubChem CID 7131132
IUPAC Name 2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC42201CB
SDS View Documents
Thermo Scientific Chemicals
CC42201CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC42201DA
SDS View Documents
Thermo Scientific Chemicals
CC42201DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

32002-72-5
218.25
APXSARIYDLVPAY-UHFFFAOYSA-M
7131132
CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1
C11H8NO2S
MFCD07348569
2-methyl-4-phenylthiazole-5-carboxylic acid, 2-methy-4-phenylthiazol-5-ylcarboxylic acid, 2-methyl-4-phenyl-thiazole-5-carboxylic acid, 2-methyl-4-phenylthiazol-5-ylcarboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl, 5-thiazolecarboxylic acid, 2-methyl-4-phenyl
2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid

Specifications

32002-72-5
Amber Glass Bottle
MFCD07348569
2-methyl-4-phenylthiazole-5-carboxylic acid, 2-methy-4-phenylthiazol-5-ylcarboxylic acid, 2-methyl-4-phenyl-thiazole-5-carboxylic acid, 2-methyl-4-phenylthiazol-5-ylcarboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl, 5-thiazolecarboxylic acid, 2-methyl-4-phenyl
CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1
218.25
219.26
2-Methyl-4-phenyl-1, 3-thiazole-5-carboxylic acid
97%
C11H8NO2S
250mg
APXSARIYDLVPAY-UHFFFAOYSA-M
2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid
7131132
97%
Videos
SDS
Documents

Documents