missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-octanone, 99%

Catalog No. aaa1995214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1995214 25 g
AAA1995206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1995214 Supplier Thermo Scientific Chemicals Supplier No. A1995214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7492-38-8 | C9H18O | 142.24 g/mol

2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 7492-38-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00048808
InChI Key ICSKJDZASFIJQK-UHFFFAOYSA-N
Synonym 2-methyl-4-octanone, 4-octanone, 2-methyl
PubChem CID 522566
IUPAC Name 2-methyloctan-4-one
SMILES CCCCC(=O)CC(C)C

Specifications

Boiling Point 180°C
Refractive Index 1.415
Quantity 25 g
Formula Weight 142.24
Percent Purity 99%
Chemical Name or Material 2-Methyl-4-octanone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only