missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4'-(methylthio)-2-morpholinopropiophenone 98.0+%, TCI America™
$93.21 - $695.29
Chemical Identifiers
| CAS | 71868-10-5 |
|---|---|
| Molecular Formula | C15H21NO2S |
| Molecular Weight (g/mol) | 279.398 |
| MDL Number | MFCD00083014 |
| InChI Key | LWRBVKNFOYUCNP-UHFFFAOYSA-N |
| Synonym | 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholino-1-propanone |
| PubChem CID | 92387 |
| IUPAC Name | 2-methyl-1-(4-methylsulfanylphenyl)-2-morpholin-4-ylpropan-1-one |
| SMILES | CC(C)(C(=O)C1=CC=C(C=C1)SC)N2CCOCC2 |
Chemical Identifiers
| 71868-10-5 | |
| 279.398 | |
| LWRBVKNFOYUCNP-UHFFFAOYSA-N | |
| 92387 | |
| CC(C)(C(=O)C1=CC=C(C=C1)SC)N2CCOCC2 |
| C15H21NO2S | |
| MFCD00083014 | |
| 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholino-1-propanone | |
| 2-methyl-1-(4-methylsulfanylphenyl)-2-morpholin-4-ylpropan-1-one |
Specifications
| 71868-10-5 | |
| White-Yellow | |
| C15H21NO2S | |
| 25 g | |
| 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholino-1-propanone | |
| CC(C)(C(=O)C1=CC=C(C=C1)SC)N2CCOCC2 | |
| 279.398 | |
| 279.40 | |
| Crystalline Powder |
| 75°C | |
| 210°C | |
| MFCD00083014 | |
| 3077 | |
| LWRBVKNFOYUCNP-UHFFFAOYSA-N | |
| 2-methyl-1-(4-methylsulfanylphenyl)-2-morpholin-4-ylpropan-1-one | |
| 92387 | |
| ≥98.0% (GC,T) | |
| 2-Methyl-4′-(methylthio)-2-morpholinopropiophenone |
Safety and Handling
EINECSNumber : (5)-5819
TSCA : Yes