missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4'-(methylthio)-2-morpholinopropiophenone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 75°C |
| Color | White-Yellow |
| Boiling Point | 210°C |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 279.40 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Methyl-4′-(methylthio)-2-morpholinopropiophenone |
Safety and Handling
| EINECSNumber | (5)-5819 |
| TSCA | Yes |