missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-isothiazolin-3-one, 50% aqueous solution

Catalog No. AC459970050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459970050 5 g
AC459970250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459970050 Supplier Thermo Scientific Chemicals Supplier No. 459970050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2682-20-4 | C4H5NOS | 115.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2682-20-4
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
InChI Key BEGLCMHJXHIJLR-UHFFFAOYSA-N
Synonym 2-methyl-4-isothiazolin-3-one, methylisothiazolinone, 2-methyl-4-isothiazoline-3-one, 2-methylisothiazol-3 2h-one, 2-methyl-3 2h-isothiazolone, n-methylisothiazolone, 3 2h-isothiazolone, 2-methyl, caswell no. 572a, 2-methyl-3-isothiazolone, unii-229d0e1qfa
PubChem CID 39800
ChEBI CHEBI:53620
IUPAC Name 2-methyl-1,2-thiazol-3-one
SMILES CN1C(=O)C=CS1

Specifications

Density 1.1600g/mL
pH 5.0 to 7.0 (1% aq. soln.)
Quantity 5 g
Specific Gravity 1.16
Formula Weight 115.15
Chemical Name or Material 2-Methyl-4-isothiazolin-3-one
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic if inhaled.
May cause respiratory irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 220-239-6

RUO – Research Use Only