missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4,6-bis(trichloromethyl)-1,3,5-triazine 98.0+%, TCI America™

Catalog No. M21375G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M21375G Supplier TCI America Supplier No. M21375G
Only null left

Chemical Identifiers

CAS 949-42-8
Molecular Formula C6H3Cl6N3
Molecular Weight (g/mol) 329.81
MDL Number MFCD00023178
InChI Key LETDRANQSOEVCX-UHFFFAOYSA-N
PubChem CID 13703
IUPAC Name 2-methyl-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES CC1=NC(=NC(=N1)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 5 g
Formula Weight 329.81
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4,6-bis(trichloromethyl)-1,3,5-triazine