missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4(5)-nitroimidazole, 99%

Catalog No. AAA1256930
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1256930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1256930 Supplier Thermo Scientific Chemicals Supplier No. A1256930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 696-23-1 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 696-23-1
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00005191
InChI Key FFYTTYVSDVWNMY-UHFFFAOYSA-N
Synonym 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
PubChem CID 12760
IUPAC Name 2-methyl-5-nitro-1H-imidazole
SMILES CC1=NC=C(N1)[N+]([O-])=O

Specifications

Melting Point 251°C to 254°C
Quantity 250 g
Beilstein 4032
Formula Weight 127.1
Percent Purity 99%
Chemical Name or Material 2-Methyl-4(5)-nitroimidazole
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 211-790-3
RTECSNumber NI7550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only