missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4,5-diphenylthiazole 97.0+%, TCI America™

Catalog No. M17055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17055G Supplier TCI America Supplier No. M17055G
Only null left

Chemical Identifiers

CAS 3755-83-7
Molecular Formula C16H13NS
Molecular Weight (g/mol) 251.35
MDL Number MFCD00186396
InChI Key ZFYWCVZNRMSXBT-UHFFFAOYSA-N
Synonym 2-methyl-4,5-diphenylthiazole, 4,5-diphenyl-2-methylthiazole, 2-methyl-4,5-diphenyl-thiazole, acmc-1aek0, thiazole,2-methyl-4,5-diphenyl, thiazole, 2-methyl-4,5-diphenyl
PubChem CID 77365
IUPAC Name 2-methyl-4,5-diphenyl-1,3-thiazole
SMILES CC1=NC(=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 52°C
Color Yellow
Quantity 5 g
Formula Weight 251.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4,5-diphenylthiazole
EINECSNumber (5)-0210
TSCA No