missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4(1H)-quinazolinone 98.0+%, TCI America™
$209.84 - $209.84
Chemical Identifiers
| CAS | 1769-24-0 |
|---|---|
| Molecular Formula | C9H8N2O |
| Molecular Weight (g/mol) | 160.18 |
| MDL Number | MFCD00006887,MFCD05270928 |
| InChI Key | FIEYHAAMDAPVCH-UHFFFAOYSA-N |
| PubChem CID | 15674 |
| IUPAC Name | 2-methyl-1,4-dihydroquinazolin-4-one |
| SMILES | CC1=NC(=O)C2=CC=CC=C2N1 |
Chemical Identifiers
| 1769-24-0 | |
| 160.18 | |
| FIEYHAAMDAPVCH-UHFFFAOYSA-N | |
| 2-methyl-1,4-dihydroquinazolin-4-one |
| C9H8N2O | |
| MFCD00006887,MFCD05270928 | |
| 15674 | |
| CC1=NC(=O)C2=CC=CC=C2N1 |
Specifications
| 1769-24-0 | |
| White-Yellow | |
| MFCD00006887,MFCD05270928 | |
| FIEYHAAMDAPVCH-UHFFFAOYSA-N | |
| 2-methyl-1,4-dihydroquinazolin-4-one | |
| 15674 | |
| ≥98.0% (GC,T) | |
| 2-Methyl-4(1H)-quinazolinone |
| 240°C | |
| C9H8N2O | |
| 5 g | |
| CC1=NC(=O)C2=CC=CC=C2N1 | |
| 160.18 | |
| 160.18 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : VA3677000
TSCA : No