missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-isopropoxypyrazine 97.0+%, TCI America™

Catalog No. p-7132374
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08081G 1 g
M08085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08081G Supplier TCI America Supplier No. M08081G
Only null left

Chemical Identifiers

CAS 94089-22-2
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD00059772
InChI Key FRCMVXLVVCXOHO-UHFFFAOYSA-N
Synonym 2-Isopropoxy-3-methylpyrazine
PubChem CID 3086387
IUPAC Name 2-methyl-3-propan-2-yloxypyrazine
SMILES CC1=NC=CN=C1OC(C)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 152.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-isopropoxypyrazine