missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-butyn-2-ol 98.0+%, TCI America™

Catalog No. M018025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M018025ML 25 mL
M0180500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M018025ML Supplier TCI America Supplier No. M018025ML

Chemical Identifiers

CAS 115-19-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00004467
InChI Key CEBKHWWANWSNTI-UHFFFAOYSA-N
Synonym 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol
PubChem CID 8258
IUPAC Name 2-methylbut-3-yn-2-ol
SMILES CC(C)(C#C)O

Specifications

Color Yellow
Boiling Point 103°C
Quantity 25 mL
UN Number 1987
Formula Weight 84.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-butyn-2-ol
EINECSNumber (2)-2396&(2)-3457
RTECSNumber ES0810000
TSCA Yes