Learn More
2-Methyl-3-butyn-2-ol, 98%
CAS: 115-19-5 | C5H8O | 84.12 g/mol
Supplier: Thermo Scientific Chemicals 156820025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methyl-3-butyn-2-ol | |
| 3.0°C | |
| 0.8608g/mL | |
| 21°C | |
| 1% max. tetrahydrofuran (GC) | |
| C5H8O | |
| HC≡CC(CH3)2OH | |
| 01,I,235 | |
| 0.8608 | |
| 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol | |
| CEBKHWWANWSNTI-UHFFFAOYSA-N | |
| 2-methylbut-3-yn-2-ol | |
| 8258 | |
| 84.12 | |
| Crystals or Powder |
| 115-19-5 | |
| Brown to White | |
| 104.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4200 to 1.4220 | |
| 2.5 L | |
| 01,674 | |
| 15,6108 | |
| Solubility in water: miscible. Other solubilities: miscible with acetone,benzene,cellosolve,,carbon tetrachloride,cyclohexanone,diethylene,glycol,ethyl acetate,kerosine,methyl ethyl,ketone,mineral spirits,monoethanolamine | |
| CC(C)(C#C)O | |
| 84.12 | |
| 3.02 mPa.s (20°C) | |
| 98% |
Chemical Identifiers
| 115-19-5 | |
| 84.12 | |
| 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol | |
| 2-methylbut-3-yn-2-ol |
| C5H8O | |
| CEBKHWWANWSNTI-UHFFFAOYSA-N | |
| 8258 | |
| CC(C)(C#C)O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Highly flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not eat,drink or smoke when us
GHS Signal Word: Danger
EINECSNumber : 204-070-5
RUO – Research Use Only