missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-butyn-2-ol, 98%

Catalog No. p-4510327
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC156822500 250 mL Glass bottle
AC156820010 1 L Glass bottle
AC156820025 2.5 L Glass bottle
3 options

Catalog No. AC156822500

Supplier: Thermo Scientific Chemicals 156822500

Only null left

CAS: 115-19-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115-19-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
InChI Key CEBKHWWANWSNTI-UHFFFAOYSA-N
Synonym 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol
PubChem CID 8258
IUPAC Name 2-methylbut-3-yn-2-ol
SMILES CC(C)(C#C)O

Specifications

Melting Point 3.0°C
Color Brown to White
Density 0.8608g/mL
Boiling Point 104.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. tetrahydrofuran (GC)
Packaging Glass bottle
Refractive Index 1.4200 to 1.4220
Linear Formula HC≡CC(CH3)2OH
Quantity 250 mL
Beilstein 01,I,235
Fieser 01,674
Merck Index 15,6108
Specific Gravity 0.8608
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetone,benzene,cellosolve,,carbon tetrachloride,cyclohexanone,diethylene,glycol,ethyl acetate,kerosine,methyl ethyl,ketone,mineral spirits,monoethanolamine
Viscosity 3.02 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 2-Methyl-3-butyn-2-ol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Highly flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not eat,drink or smoke when us
EINECSNumber 204-070-5

RUO – Research Use Only