missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Methyl-3-butyn-2-ol

Catalog No. 1297631L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1297631L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1297631L Supplier Sigma Aldrich Supplier No. 1297631L

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methyl-3-butyn-2-ol

Specifications

CAS 115-19-5
Melting Point 2.6°C (lit.)
Density 0.868 g/mL (at 25°C (literature))
Boiling Point 104°C (lit.)
Molecular Formula C5H8O
Refractive Index n20/D 1.42 (literature)
Linear Formula HC///CC(CH3)2 OH
MDL Number MFCD00004467
Quantity 1 L
Synonym Dimethyl ethynyl carbinol
Molecular Weight (g/mol) 84.12
Percent Purity 98%
Show More Show Less