missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-buten-2-ol 97.0+%, TCI America™
$90.48 - $415.84
Chemical Identifiers
| CAS | 115-18-4 |
|---|---|
| Molecular Formula | C5H10O |
| Molecular Weight (g/mol) | 86.134 |
| MDL Number | MFCD00004470 |
| InChI Key | HNVRRHSXBLFLIG-UHFFFAOYSA-N |
| Synonym | 2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol |
| PubChem CID | 8257 |
| IUPAC Name | 2-methylbut-3-en-2-ol |
| SMILES | CC(C)(C=C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M017825ML
|
TCI America
M017825ML |
25 mL |
Each for $90.48
|
|
|||||
|
M0178500ML
|
TCI America
M0178500ML |
500 mL |
Each for $415.84
|
|
|||||
Chemical Identifiers
| 115-18-4 | |
| 86.134 | |
| HNVRRHSXBLFLIG-UHFFFAOYSA-N | |
| 8257 | |
| CC(C)(C=C)O |
| C5H10O | |
| MFCD00004470 | |
| 2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol | |
| 2-methylbut-3-en-2-ol |
Specifications
| 115-18-4 | |
| Yellow | |
| MFCD00004470 | |
| 1987 | |
| HNVRRHSXBLFLIG-UHFFFAOYSA-N | |
| 2-methylbut-3-en-2-ol | |
| 8257 | |
| ≥97.0% (GC) | |
| 2-Methyl-3-buten-2-ol |
| -28°C | |
| C5H10O | |
| 25 mL | |
| 2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol | |
| CC(C)(C=C)O | |
| 86.134 | |
| 86.13 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2393
RTECSNumber : EM9472000
TSCA : Yes