missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-buten-2-ol 97.0+%, TCI America™

Catalog No. M017825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M017825ML 25 mL
M0178500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M017825ML Supplier TCI America Supplier No. M017825ML

Chemical Identifiers

CAS 115-18-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00004470
InChI Key HNVRRHSXBLFLIG-UHFFFAOYSA-N
Synonym 2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
PubChem CID 8257
IUPAC Name 2-methylbut-3-en-2-ol
SMILES CC(C)(C=C)O

Specifications

Melting Point -28°C
Color Yellow
Quantity 25 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-buten-2-ol
EINECSNumber (2)-2393
RTECSNumber EM9472000
TSCA Yes