missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-buten-1-ol, 98%

Catalog No. AC425690010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425690010 1 g Glass bottle
AC425690050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425690010 Supplier Thermo Scientific Chemicals Supplier No. 425690010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4516-90-9 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4516-90-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
InChI Key NVGOATMUHKIQQG-UHFFFAOYSA-N
Synonym 2-methyl-3-buten-1-ol, 3-buten-1-ol, 2-methyl, 2-methyl-but-3-en-1-ol, 3-buten-1-ol, 2-methyl-, 2s, acmc-20mpc0, acmc-1bn7w, 2-methyl-3-butene-1-ol, 1-hydroxy-2-methyl-3-butene, nvgoatmuhkiqqg-uhfffaoysa
PubChem CID 94292
IUPAC Name 2-methylbut-3-en-1-ol
SMILES CC(CO)C=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8350g/mL
Boiling Point 120.0°C to 121.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH(CH3)CH2OH
Refractive Index 1.4260 to 1.4280 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.835
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-buten-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Recommended Storage Refrigerator +4°C

RUO – Research Use Only